Learn More
Biomatik Corporation GABARAP anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07105
Encompass_Preferred
The GABARAP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Gamma-aminobutyric acid receptor-associated protein (2-117AA). Applications: ELISA, IF. Recommended dilution: IF:1:100-200. UniProt: O95166. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6692