Learn More
Biomatik Corporation CRP anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506695
Encompass_Preferred
The CRP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human C-reactive protein (19-224AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: CRP PTX1. UniProt: P02741. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6695 Supplier Biomatik Corporation Supplier No. CAC07108
The CRP Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human C-reactive protein (19-224AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: CRP PTX1. UniProt: P02741. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.