Learn More
Biomatik Corporation ISG15 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506697
Encompass_Preferred
Isg15 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ISG15. The Isg15 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ubiquitin-Like Protein Isg15 Protein (2-157Aa). Applications: ELISA, WB, IP. Target Synonyms: ISG15. Accession Number: P05161. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6697 Supplier Biomatik Corporation Supplier No. CAC07110
Isg15 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ISG15. The Isg15 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ubiquitin-Like Protein Isg15 Protein (2-157Aa). Applications: ELISA, WB, IP. Target Synonyms: ISG15. Accession Number: P05161. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.