Learn More
Biomatik Corporation MKI67 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07112
Encompass_Preferred
Mki67 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MKI67. The Mki67 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Proliferation Marker Protein Ki-67 Protein (3120-3256Aa). Applications: ELISA, IHC. Target Synonyms: MKI67. Accession Number: P46013. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6699