Learn More
Biomatik Corporation ANXA5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506701
Encompass_Preferred
The ANXA5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Annexin A5 protein (2-320AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:200-1000, IHC:1:20-1:200. Alternative Names: Anchorin CII, Annexin V, Annexin-5, Calphobindin I, Endonexin II, Lipocortin V, Placental anticoagulant protein 4, Placental anticoagulant protein I, Thromboplastin inhibitor, Vascular anticoagulant-alpha, ANX5, ENX2, PP4, ANXA5. UniProt: P08758. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6701 Supplier Biomatik Corporation Supplier No. CAC07114
The ANXA5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Annexin A5 protein (2-320AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:200-1000, IHC:1:20-1:200. Alternative Names: Anchorin CII, Annexin V, Annexin-5, Calphobindin I, Endonexin II, Lipocortin V, Placental anticoagulant protein 4, Placental anticoagulant protein I, Thromboplastin inhibitor, Vascular anticoagulant-alpha, ANX5, ENX2, PP4, ANXA5. UniProt: P08758. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.