Learn More
Biomatik Corporation COL4A1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506706
Encompass_Preferred
The COL4A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Collagen alpha-1 (IV) chain protein (30-167AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Procollagen-proline, 2-oxoglutarate-4-dioxygenase subunit alpha-1;P4HA;P4HA1. UniProt: P02462. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6706 Supplier Biomatik Corporation Supplier No. CAC07119
The COL4A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Collagen alpha-1 (IV) chain protein (30-167AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Procollagen-proline, 2-oxoglutarate-4-dioxygenase subunit alpha-1;P4HA;P4HA1. UniProt: P02462. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.