Learn More
Biomatik Corporation BMP3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506709
Encompass_Preferred
The BMP3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Bone morphogenetic protein 3 protein (363-472AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Bone morphogenetic protein 3A, BMP3, BMP3A. UniProt: P12645. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6709 Supplier Biomatik Corporation Supplier No. CAC07122
The BMP3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Bone morphogenetic protein 3 protein (363-472AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Bone morphogenetic protein 3A, BMP3, BMP3A. UniProt: P12645. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.