Learn More
Biomatik Corporation DCX anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07130
Encompass_Preferred
The DCX Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Neuronal migration protein doublecortin protein (1-441AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Neuronal migration protein doublecortin, DCX, DBCN, LISX, Doublin, Lissencephalin-X, Lis-X. UniProt: O43602. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6717