Learn More
Biomatik Corporation AGO2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07145
Encompass_Preferred
The AGO2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Protein argonaute-2 protein (517-818AA). Applications: ELISA, WB, IHC, ChIP. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Argonaute RISC catalytic component 2, Eukaryotic translation initiation factor 2C 2, PAZ Piwi domain protein, Protein slicer, EIF2C2, AGO2. UniProt: Q9UKV8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6732