Learn More
Biomatik Corporation HSP90AB1 anti-Human, Mouse, Rat, Zebrafish, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07147
Encompass_Preferred
The HSP90AB1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat, Zebrafish. Immunogen: Recombinant human human Heat shock protein HSP 90-beta protein (150-355AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-1:5000, IHC:1:100-1:500, IF:1:50-1:500. Alternative Names: HSP 90, Heat shock 84 kDa, HSP 84, HSP90AB1, HSP90B, HSPC2, HSPCB. UniProt: P08238. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6734