Learn More
Biomatik Corporation ALDH3A1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07162
Encompass_Preferred
The ALDH3A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Aldehyde dehydrogenase, dimeric NADP-preferring protein (210-453AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:200-1000, IHC:1:20-1:200. Alternative Names: Aldehyde dehydrogenase, dimeric NADP-preferring, ALDHIII, Aldehyde dehydrogenase 3, Aldehyde dehydrogenase family 3 member A1, ALDH3A1, ALDH3. UniProt: P30838. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6749