Learn More
Biomatik Corporation YWHAB anti-Human, Mouse, Rat, Zebrafish, Rabbit Polyclonal Antibody, 100UL
Supplier: Biomatik Corporation CAC07166
Ywhab Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to YWHAB. The Ywhab Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 14-3-3 Protein Beta; Alpha Protein (1-246Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: YWHAB. Accession Number: P31946. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.