Learn More
Biomatik Corporation MMP7 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506760
Encompass_Preferred
Mmp7 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MMP7. The Mmp7 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Matrilysin Protein (95-267Aa). Applications: ELISA, IHC. Target Synonyms: MMP7. Accession Number: P09237. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6760 Supplier Biomatik Corporation Supplier No. CAC07173
Mmp7 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MMP7. The Mmp7 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Matrilysin Protein (95-267Aa). Applications: ELISA, IHC. Target Synonyms: MMP7. Accession Number: P09237. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.