Learn More
Biomatik Corporation DNMT3B anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506763
Encompass_Preferred
The DNMT3B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human DNA (cytosine-5)-methyltransferase 3B protein (1-100AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, M.HsaIIIB, DNMT3B. UniProt: Q9UBC3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6763 Supplier Biomatik Corporation Supplier No. CAC07176
The DNMT3B Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human DNA (cytosine-5)-methyltransferase 3B protein (1-100AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: DNA (cytosine-5)-methyltransferase 3B, DNA methyltransferase HsaIIIB, DNA MTase HsaIIIB, M.HsaIIIB, DNMT3B. UniProt: Q9UBC3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.