Learn More
Biomatik Corporation TRIM28 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506776
Encompass_Preferred
Trim28 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRIM28. The Trim28 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transcription Intermediary Factor 1-Beta Protein (22-291Aa). Applications: ELISA, WB, IHC, IP. Target Synonyms: TRIM28. Accession Number: Q13263. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6776 Supplier Biomatik Corporation Supplier No. CAC07189
Trim28 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRIM28. The Trim28 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transcription Intermediary Factor 1-Beta Protein (22-291Aa). Applications: ELISA, WB, IHC, IP. Target Synonyms: TRIM28. Accession Number: Q13263. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.