Learn More
Biomatik Corporation INHBA anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506778
Encompass_Preferred
Inhba Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to INHBA. The Inhba Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Inhibin Beta A Chain Protein (311-426Aa). Applications: ELISA, WB, IHC. Target Synonyms: INHBA. Accession Number: P08476. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6778 Supplier Biomatik Corporation Supplier No. CAC07191
Inhba Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to INHBA. The Inhba Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Inhibin Beta A Chain Protein (311-426Aa). Applications: ELISA, WB, IHC. Target Synonyms: INHBA. Accession Number: P08476. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.