Learn More
Biomatik Corporation CDH12 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506779
Encompass_Preferred
The CDH12 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cadherin-12 protein (56-605AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Brain cadherin, BR-cadherin, Neural type cadherin 2, N-cadherin 2, CDH12. UniProt: P55289. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6779 Supplier Biomatik Corporation Supplier No. CAC07192
The CDH12 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cadherin-12 protein (56-605AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Brain cadherin, BR-cadherin, Neural type cadherin 2, N-cadherin 2, CDH12. UniProt: P55289. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.