Learn More
Biomatik Corporation LEF1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07199
Encompass_Preferred
Lef1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LEF1. The Lef1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lymphoid Enhancer-Binding Factor 1 Protein (75-216Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: LEF1. Accession Number: Q9UJU2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6786