Learn More
Biomatik Corporation COL4A3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07203
Encompass_Preferred
The COL4A3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Collagen alpha-3 (IV) chain protein (1446-1667AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Goodpasture antigen, COL4A3. UniProt: Q01955. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6790