Learn More
Biomatik Corporation CYP11A1 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506792
Encompass_Preferred
The CYP11A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Cholesterol side-chain cleavage enzyme, mitochondrial protein (40-521AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: CYPXIA1, Cholesterol desmolase, Cytochrome P450 11A1, Cytochrome P450 (scc), CYP11A1, CYP11A. UniProt: P05108. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6792 Supplier Biomatik Corporation Supplier No. CAC07205
The CYP11A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant human Cholesterol side-chain cleavage enzyme, mitochondrial protein (40-521AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: CYPXIA1, Cholesterol desmolase, Cytochrome P450 11A1, Cytochrome P450 (scc), CYP11A1, CYP11A. UniProt: P05108. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.