Learn More
Biomatik Corporation S100A9 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07212
Encompass_Preferred
S100A9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to S100A9. The S100A9 Polyclonal Antibody has been validated for the following applications: ELISA, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein S100-A9 Protein (2-114Aa). Applications: ELISA, IF. Target Synonyms: S100A9. Accession Number: P06702. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6799