Learn More
Biomatik Corporation POSTN anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07219
Encompass_Preferred
Postn Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to POSTN. The Postn Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Periostin Protein (24-836Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: POSTN. Accession Number: Q15063. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6806