Learn More
Biomatik Corporation AQP1 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07222
Encompass_Preferred
The AQP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Aquaporin-1 protein (220-269AA). Applications: ELISA, WB, IF. Recommended dilution: WB: 1:1000-1:5000, IF:1:50-1:200. Alternative Names: Aquaporin-CHIP, Urine water channel, Water channel protein for red blood cells and kidney proximal tubule, AQP1, CHIP28. UniProt: P29972. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6809