Learn More
Biomatik Corporation fbpA anti-Mycobacterium tuberculosis, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506810
Encompass_Preferred
The fbpA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mycobacterium tuberculosis. Immunogen: Recombinant Mycobacterium tuberculosis Antigen 85-A protein (53-331AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: Antigen 85 complex A, Fibronectin-binding protein A Mycolyl transferase 85A, fbpA, mpt44. UniProt: P9WQP3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6810 Supplier Biomatik Corporation Supplier No. CAC07223
The fbpA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mycobacterium tuberculosis. Immunogen: Recombinant Mycobacterium tuberculosis Antigen 85-A protein (53-331AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: Antigen 85 complex A, Fibronectin-binding protein A Mycolyl transferase 85A, fbpA, mpt44. UniProt: P9WQP3. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.