Learn More
Biomatik Corporation ERGIC3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506812
Encompass_Preferred
The ERGIC3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Endoplasmic reticulum-Golgi intermediate compartment protein 3 protein (47-341AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Serologically defined breast cancer antigen NY-BR-84, ERGIC3, C20orf47, ERV46, SDBCAG84, CGI-54, PRO0989. UniProt: Q9Y282. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6812 Supplier Biomatik Corporation Supplier No. CAC07225
The ERGIC3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Endoplasmic reticulum-Golgi intermediate compartment protein 3 protein (47-341AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Serologically defined breast cancer antigen NY-BR-84, ERGIC3, C20orf47, ERV46, SDBCAG84, CGI-54, PRO0989. UniProt: Q9Y282. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.