Learn More
Biomatik Corporation BCAT2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506815
Encompass_Preferred
The BCAT2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Branched-chain-amino-acid aminotransferase, mitochondrial (28-392AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Branched-chain-amino-acid aminotransferase, mitochondrial, Placental protein 18, BCATM, BCT2, ECA40, BCAT2. UniProt: O15382. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6815 Supplier Biomatik Corporation Supplier No. CAC07228
The BCAT2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Branched-chain-amino-acid aminotransferase, mitochondrial (28-392AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Branched-chain-amino-acid aminotransferase, mitochondrial, Placental protein 18, BCATM, BCT2, ECA40, BCAT2. UniProt: O15382. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.