Learn More
Biomatik Corporation Hemoglobin anti-Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07231
Encompass_Preferred
The Hemoglobin Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: Mouse Hemoglobin protein (Native Protein). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6818