Learn More
Biomatik Corporation FFAR3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07234
Encompass_Preferred
The FFAR3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Free fatty acid receptor 3 protein (280-346AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Free fatty acid receptor 3, G-protein coupled receptor 41, FFAR3, GPR41. UniProt: O14843. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6821