Learn More
Biomatik Corporation FGFR3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07235
Encompass_Preferred
The FGFR3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Fibroblast growth factor receptor 3 protein (23-375AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:100-1:500, IF:1:50-1:500. Alternative Names: CD_antigen=CD333 FGFR3 JTK4. UniProt: P22607. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6822