Learn More
Biomatik Corporation HNRNPA1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07236
Encompass_Preferred
The HNRNPA1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Heterogeneous nuclear ribonucleoprotein A1 protein (2-354AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Helix-destabilizing protein, Single-strand RNA-binding protein, hnRNP core protein A1, HNRNPA1, HNRPA1. UniProt: P09651. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6823