Learn More
Biomatik Corporation Trem2 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506828
Encompass_Preferred
Trem2 Polyclonal Antibody is a Mus musculus (Mouse (Mus musculus)) Polyclonal Antibody to Trem2. The Trem2 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Mus musculus (Mouse (Mus musculus)). Immunogen: Recombinant Mouse Triggering Receptor Expressed On Myeloid Cells 2 Protein (19-171Aa). Applications: ELISA, WB. Target Synonyms: Trem2. Accession Number: Q99NH8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6828 Supplier Biomatik Corporation Supplier No. CAC07241
Trem2 Polyclonal Antibody is a Mus musculus (Mouse (Mus musculus)) Polyclonal Antibody to Trem2. The Trem2 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Mus musculus (Mouse (Mus musculus)). Immunogen: Recombinant Mouse Triggering Receptor Expressed On Myeloid Cells 2 Protein (19-171Aa). Applications: ELISA, WB. Target Synonyms: Trem2. Accession Number: Q99NH8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.