Learn More
Biomatik Corporation IL1RN anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506833
Encompass_Preferred
Il1Rn Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IL1RN. The Il1Rn Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-1 Receptor Antagonist Protein (26-177Aa). Applications: ELISA, WB. Target Synonyms: IL1RN. Accession Number: P18510. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6833 Supplier Biomatik Corporation Supplier No. CAC07246
Il1Rn Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IL1RN. The Il1Rn Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interleukin-1 Receptor Antagonist Protein (26-177Aa). Applications: ELISA, WB. Target Synonyms: IL1RN. Accession Number: P18510. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.