Learn More
Biomatik Corporation CALR anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07249
Encompass_Preferred
The CALR Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Calreticulin protein (18-417AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Calreticulin, CRP55, Calregulin, Endoplasmic reticulum resident protein 60, ERp60, HACBP, CALR, CRTC. UniProt: P27797. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6836