Learn More
Biomatik Corporation TERT anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07250
Encompass_Preferred
Tert Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TERT. The Tert Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Telomerase Reverse Transcriptase Protein (281-436Aa). Applications: ELISA, IHC. Target Synonyms: TERT. Accession Number: O14746. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6837