Learn More
Biomatik Corporation TGM2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506838
Encompass_Preferred
Tgm2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TGM2. The Tgm2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein-Glutamine Gamma-Glutamyltransferase 2 Protein (4-544Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TGM2. Accession Number: P21980. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6838 Supplier Biomatik Corporation Supplier No. CAC07251
Tgm2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TGM2. The Tgm2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein-Glutamine Gamma-Glutamyltransferase 2 Protein (4-544Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TGM2. Accession Number: P21980. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.