Learn More
Biomatik Corporation CD59 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07258
Encompass_Preferred
The CD59 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human CD59 glycoprotein (26-102AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: 1F5 antigen 20 kDa homologous restriction factor MAC-inhibitory protein MEM43 antigen Membrane attack complex inhibition factor Membrane inhibitor of reactive lysis Protectin CD_antigen=CD59 CD59 MIC11, MIN1, MIN2, MIN3, MSK21. UniProt: P13987. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6845