Learn More
Biomatik Corporation DNMT3A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07259
Encompass_Preferred
The DNMT3A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human DNA (cytosine-5)-methyltransferase 3A protein (680-902AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: DNA methyltransferase HsaIIIA, DNMT3A. UniProt: Q9Y6K1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6846