Learn More
Biomatik Corporation ANXA6 anti-Human, Rat, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506848
Encompass_Preferred
The ANXA6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant Human Annexin A6 protein (2-245AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: 67 kDa calelectrin, Annexin VI, Annexin-6, Calphobindin-II, Chromobindin-20, Lipocortin VI, Protein III, p68, p70, ANXA6, ANX6. UniProt: P08133. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6848 Supplier Biomatik Corporation Supplier No. CAC07261
The ANXA6 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Rat. Immunogen: Recombinant Human Annexin A6 protein (2-245AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: 67 kDa calelectrin, Annexin VI, Annexin-6, Calphobindin-II, Chromobindin-20, Lipocortin VI, Protein III, p68, p70, ANXA6, ANX6. UniProt: P08133. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.