Learn More
Biomatik Corporation RORC anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07267
Encompass_Preferred
Rorc Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RORC. The Rorc Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nuclear Receptor Ror-Gamma Protein (1-518Aa). Applications: ELISA, WB, IHC. Target Synonyms: RORC. Accession Number: P51449. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6854