Learn More
Biomatik Corporation FASN anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506871
Encompass_Preferred
The FASN Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Fatty acid synthase protein (2155-2495AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: FASN, FAS. UniProt: P49327. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6871 Supplier Biomatik Corporation Supplier No. CAC07284
The FASN Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Fatty acid synthase protein (2155-2495AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: FASN, FAS. UniProt: P49327. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.