Learn More
Biomatik Corporation mpt64 anti-Mycobacterium tuberculosis, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07286
Encompass_Preferred
Mpt64 Polyclonal Antibody is a Mycobacterium tuberculosis Polyclonal Antibody to mpt64. The Mpt64 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Mycobacterium tuberculosis. Immunogen: Recombinant Mycobacterium Tuberculosis Immunogenic Protein Mpt64 Protein (24-228Aa). Applications: ELISA, WB. Target Synonyms: mpt64. Accession Number: P9WIN9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6873