Learn More
Biomatik Corporation CA2 anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07295
Encompass_Preferred
The CA2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant human Carbonic anhydrase 2 protein (2-260AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Carbonate dehydratase II, Carbonic anhydrase C, CAC, Carbonic anhydrase II, CA-II, CA2. UniProt: P00918. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6882