Learn More
Biomatik Corporation AIF1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506885
Encompass_Preferred
The AIF1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Allograft inflammatory factor 1 protein (2-147AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: AIF-1, Ionized calcium-binding adapter molecule 1, Protein G1, AIF1, G1, IBA1. UniProt: P55008. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6885 Supplier Biomatik Corporation Supplier No. CAC07299
The AIF1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Allograft inflammatory factor 1 protein (2-147AA). Applications: ELISA, IHC, IF. Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: AIF-1, Ionized calcium-binding adapter molecule 1, Protein G1, AIF1, G1, IBA1. UniProt: P55008. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.