Learn More
Biomatik Corporation FABP1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07300
Encompass_Preferred
The FABP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Fatty acid-binding protein, liver protein (1-127AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: Fatty acid-binding protein 1 Liver-type fatty acid-binding protein FABP1 FABPL. UniProt: P07148. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6886