Learn More
Biomatik Corporation TFRC anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07319
Encompass_Preferred
Tfrc Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TFRC. The Tfrc Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transferrin Receptor Protein 1 Protein (94-754Aa). Applications: ELISA, IHC, IF. Target Synonyms: TFRC. Accession Number: P02786. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6905