Learn More
Biomatik Corporation rpsB anti-Escherichia coli, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506906
Encompass_Preferred
Rpsb Polyclonal Antibody is a Escherichia coli Polyclonal Antibody to rpsB. The Rpsb Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli. Immunogen: Recombinant Escherichia Coli 30S Ribosomal Protein S2 Protein (2-241Aa). Applications: ELISA, WB. Target Synonyms: rpsB. Accession Number: P0A7V0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6906 Supplier Biomatik Corporation Supplier No. CAC07320
Rpsb Polyclonal Antibody is a Escherichia coli Polyclonal Antibody to rpsB. The Rpsb Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli. Immunogen: Recombinant Escherichia Coli 30S Ribosomal Protein S2 Protein (2-241Aa). Applications: ELISA, WB. Target Synonyms: rpsB. Accession Number: P0A7V0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.