Learn More
Biomatik Corporation SERPINB3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501506909
Encompass_Preferred
Serpinb3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SERPINB3. The Serpinb3 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serpin B3 Protein (2-383Aa). Applications: ELISA, IHC, IF. Target Synonyms: SERPINB3. Accession Number: P29508. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6909 Supplier Biomatik Corporation Supplier No. CAC07323
Serpinb3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SERPINB3. The Serpinb3 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serpin B3 Protein (2-383Aa). Applications: ELISA, IHC, IF. Target Synonyms: SERPINB3. Accession Number: P29508. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.