Learn More
Biomatik Corporation BSG anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07324
Encompass_Preferred
The BSG Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Basigin protein (138-321AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: 5F7, 5F7, Collagenase stimulatory factor, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor, BSG. UniProt: P35613. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-6910