Learn More
Biomatik Corporation IAPP anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07325
Encompass_Preferred
Iapp Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IAPP. The Iapp Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Islet Amyloid Polypeptide Protein (34-70Aa). Applications: ELISA, WB, IHC. Target Synonyms: IAPP. Accession Number: P10997. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6911