Learn More
Biomatik Corporation Ly6e anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07330
Encompass_Preferred
Ly6E Polyclonal Antibody is a Mus musculus (Mouse (Mus musculus)) Polyclonal Antibody to Ly6e. The Ly6E Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Mus musculus (Mouse (Mus musculus)). Immunogen: Recombinant Mouse Lymphocyte Antigen 6E Protein (21-102Aa). Applications: ELISA, WB. Target Synonyms: Ly6e. Accession Number: Q64253. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-6916